[EN]12 COMPONENTS: 10MG/L EACH IN HNO3 2 1 * 100 mL
CMPLN9300235.L1
Frequently Bought Together
Acido trifluoroacetico ≥99.9%, HiPerSolv CHROMANORM® per LC-MS
: VWR Chemicals
Ammoniaca soluzione 25%, AnalaR NORMAPUR® per analisi
: VWR Chemicals
Strisce reattive pH, non bleeding, Dosatest
: VWR Chemicals
Strisce reattive, perossidi, Dosatest
: VWR Chemicals
Acido formico ≥99%, HiPerSolv CHROMANORM® per LC-MS
: VWR Chemicals
Joint grease, K.W.S.
: POLLATH