1 component: Se 1mg/l in HNO3 2% CRM, ISO 17034 and ISO 17025 Conservation 12 months. (6 months after opening) Store under normal laboratory conditions, at temperatures between 15C to 25C 1 * 100 mL
Catalog # CMPLRM001486L1
Supplier: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together