[EN]7 COMPONENT 5MG/L EACH IN H2O 1 * 100 mL
CMPLRM008475L1
Frequently Bought Together
Ammoniaca soluzione 25%, AnalaR NORMAPUR® per analisi
: VWR Chemicals
Acido trifluoroacetico ≥99.9%, HiPerSolv CHROMANORM® per LC-MS
: VWR Chemicals
Strisce reattive pH, non bleeding, Dosatest
: VWR Chemicals
Acido formico ≥99%, HiPerSolv CHROMANORM® per LC-MS
: VWR Chemicals