Hand Scrapers
Hand scraper, red
Codice catalogo SAMP8336R-1
Nome del fornitore: Sampling Systems
Vai a
Product Details & Documents
These robust hand scrapers are perfect for scraping large areas.
- Made from high quality PP
- Robust and incorporates a large sturdy handle
- Large 250 mm wide blade
- Comes in three distinctive colours
The range of robust and sturdy hand scrapers from Sampling Systems are perfect for easily scraping large areas due to their large blade size and robust handle. The performance is further enhanced by the fact it is made out of PP giving it very good chemical resistance.
Specifiche
- Descrizione:Hand scraper, red
- Seq:02