SFr. 0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
Confirms with all Requirements of BP/EP/USP/FCC/E322(ii)/IP/JSFA/FAO/WHO CCFAS vol4 1 * 25 kg
CMPP10045.0025
:
CUSTOM MADE CHEMICALS
Confirms with all Requirements of BP/EP/USP/FCC/E322(ii)/IP/JSFA/FAO/WHO CCFAS vol4 1 * 25 kg
CMPP10045.0025
:
CUSTOM MADE CHEMICALS
Frequently Bought Together
Environmental Standards - 4 components: 1000mg/l each of Ca K Mg Na in HNO3 5% CRM, ISO 17034 and ISO 17025 Conservation 36 months. (12 months after opening) Store under normal laboratory conditions, at temperatures between 15C to 25C 1 * 100 mL
: CUSTOM MADE CHEMICALS LAB
Lovibond® Hardness Total Drop Test Kit
: Lovibond
Powder pillows, ammonia
: Hach
Reagents for Phosphate Determination using Lovibond® MD photometers
: Lovibond
Orange II ≥85 % (dye content), certified by the Biological Stain Commission, Sigma-Aldrich®
: SIGMA ALDRICH MICROSCOPY
Acid Red 183 30% (dye content), Sigma-Aldrich®
: SIGMA ALDRICH MICROSCOPY
[EN]BIOTECTOR 1.8N SULFURIC ACID 80MG/L 1 * 20 L
: Hach