FlashGel Ladefarbstoff (5 X 1ML) 1 * 5 ST
Katalognr. BZYM850462-5ML
Supplier: BIOZYM
Frequently Bought Together
VWR® Cryoboxes, Cardboard, 136×136 mm, with Full Labelling
Supplier: VWR Collection
VWR®, CryoBox, cartone, 136x136 mm
Supplier: VWR Collection
VWR®, Cryo coder cap inserts
Supplier: VWR Collection
VWR®, Flaconi per il campionamento dell'acqua
Supplier: VWR Collection
[EN]PLATE 1536W WH F-BOT HI-BASE N-BIND 1 * 60 pezzi
Supplier: Greiner Bio-One
PIASTRA PETRI Ø 90 MM VENT. 16MM STERILE 1 * 500 pezzi
Supplier: Anicrin
Inserti per colture cellulari, Nunc™
Supplier: Thermo Fisher Scientific
Cap sealing mats for VersiPlate 96-well PCR strip tube plates, VersiCap™ Mats
Supplier: Thermo Fisher Scientific
CryoBoxes with hinged lid, for cryovials, 1 to 5 ml, TruCool®
Supplier: BIOCISION
96-Well assay plates
Supplier: BIOTIX
Cryobox, colorate, Nalgene®
Supplier: Thermo Fisher Scientific
Deepwell Plates 96-Well, 2000 µl
Supplier: EPPENDORF
Peqlab, PCR Plates, 24- and 48-Well
Supplier: VWR Collection
Screw caps for internal thread sample tubes
Supplier: Simport Scientific