4-chloro-5-iodopyridin-2-amine 1 * 5 g
Catalog # PHBLPBZ0214-5G
Supplier: Pharmablock (USA)
Frequently Bought Together
2-Propanolo ≥70% (v/v), SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
1,2,3,4-Tetraidronaftalene per sintesi, Sigma-Aldrich®
Supplier: Merck
Glicerina ≥98%, GPR RECTAPUR®
Supplier: VWR Chemicals
n-Esano, EMPLURA®, Supelco®
Supplier: Merck
n-Esano
Supplier: Roth Carl
HALAG CHEMIE HALADES 191 KANISTER 10KG 1 * 10 kg
Supplier: MARKET SOURCE PART PROCESS
Clorobenzene ≥99%
Supplier: Thermo Fisher Scientific
Cicloesano ≥99.7%, CHROMASOLV™ per HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
2-Propanolo ≥99.9%, CHROMASOLV™ Plus per HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
n-Pentano, extra dry 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Etile acetato ≥99.5% ACS
Supplier: Thermo Fisher Scientific
n-Esano
Supplier: Roth Carl
Diclorometano, extra dry over Molecular Sieve 99.8% stabilizzato, AcroSeal®
Supplier: Thermo Fisher Scientific