(2S,4R)-1-[(2S)-2-(4-aminobutanoylamino)-3,3-dimethyl-butanoyl]-4-hydroxy-N-[[4-(4-methylthiazol-5-yl)phenyl]methyl]pyrrolidine-2-ca rboxamide hydrochloride 1 * 1 g
Catalog # PHBLPBY1922-1-1G
Supplier: Pharmablock (USA)
(2S,4R)-1-[(2S)-2-(4-aminobutanoylamino)-3,3-dimethyl-butanoyl]-4-hydroxy-N-[[4-(4-methylthiazol-5-yl)phenyl]methyl]pyrrolidine-2-ca rboxamide hydrochloride 1 * 1 g
Catalog # PHBLPBY1922-1-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Toluene, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
Dimetilsolfossido, anidro ≥99.8%
Supplier: Thermo Fisher Scientific
Etanolo (untaxed) ~96% (v/v), CHROMASOLV™ ACS per spettrofotometria UV extra fine spirit, E15 o 1, Riedel-de Haën™
Supplier: Honeywell Chemicals
TOLUOL CHROMASOLV FUER DI 1 * 18 l
Supplier: Honeywell Chemicals
(±)-2-Metiltetraidrofurano 99+% stabilizzato, puro
Supplier: Thermo Fisher Scientific
Tetrabutilammonio idrossido 0.4 M in water per HPLC
Supplier: Thermo Fisher Scientific
Glicerina ≥83.5-88.5% (w/w) in soluzione acquosa, puro
Supplier: Thermo Fisher Scientific
Acetonitrile 99.9%, ECD tested for residue analysis
Supplier: Thermo Fisher Scientific
Etile acetato ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS per analisi
Supplier: VWR Chemicals
ter-Butil metil etere ≥99.0%, AnalaR NORMAPUR® ACS
Supplier: VWR Chemicals