[4-[[(2S)-2-[[(2S)-2-(tert-butoxycarbonylamino)-3-methyl-butanoyl]amino]-5-ureido-pentanoyl]amino]phenyl]methyl (4-nitrophenyl) carb onate 1 * 1 g
PHBLPBY0272-1G
[4-[[(2S)-2-[[(2S)-2-(tert-butoxycarbonylamino)-3-methyl-butanoyl]amino]-5-ureido-pentanoyl]amino]phenyl]methyl (4-nitrophenyl) carb onate 1 * 1 g
PHBLPBY0272-1G
Frequently Bought Together
Glicerolo (glicerina) USP, Ph. Eur., BP
: VWR Chemicals
1,3-Diossolano ≥99.0% stabilizzato, GPR RECTAPUR® per sintesi
: VWR Chemicals
Piridina, extra dry over Molecular Sieve 99.5%, AcroSeal®
: Thermo Fisher Scientific
Carbonio solfuro 99.9% per HPLC
: Thermo Fisher Scientific
Acrilonitrile 99+%
: Thermo Fisher Scientific
Cicloesano 99+%, puro
: Thermo Fisher Scientific
FISHER CHEMICAL ACETONITRIL UHPLC GRADIENT GRAD
: MARKET SOURCE PART PROCESS
Acqua, TraceSELECT™ per analisi in tracce, Riedel-de Haën™
: Honeywell Chemicals
1-Esanolo 98%, per analisi
: Honeywell Chemicals
Piridina ≥99.0% ACS, Riedel-de Haën™
: Honeywell Chemicals