(2,5-dioxopyrrolidin-1-yl) 3-[2-[2-[2-[2-(2,5-dioxopyrrol-1-yl)ethoxy]ethoxy]ethoxy]ethoxy]propanoate 1 * 1 g
Catalog # PHBLPBY0266-1G
Anbieter: Pharmablock (USA)
(2,5-dioxopyrrolidin-1-yl) 3-[2-[2-[2-[2-(2,5-dioxopyrrol-1-yl)ethoxy]ethoxy]ethoxy]ethoxy]propanoate 1 * 1 g
Catalog # PHBLPBY0266-1G
Anbieter: Pharmablock (USA)
Frequently Bought Together
ETHYL ACETATE ANHYDROUS, 99.8 % 1 * 2 L
Anbieter: Merck
Pirogallolo per sintesi, Sigma-Aldrich®
Anbieter: Merck
2-Propanolo 99.5% per HPLC
Anbieter: Thermo Fisher Scientific
Metanolo ≥99.9%, CHROMASOLV™ per analisi GC capillare, Riedel-de Haën™
Anbieter: Honeywell Chemicals
2-Nitrophenylhydrazine, moistened 30% with water 97%
Anbieter: Thermo Fisher Scientific
Nitrometano 96% per spettroscopia
Anbieter: Thermo Fisher Scientific
Metanolo ≥99.9%, CHROMASOLV™, grado gradiente per HPLC, Riedel-de Haën™
Anbieter: Honeywell Chemicals
(±)-2-Metiltetraidrofurano, anidro stabilizzato for grignard-reaction
Anbieter: Thermo Fisher Scientific
Carbonio solfuro 99.9% per HPLC
Anbieter: Thermo Fisher Scientific
2,2,4-Trimetil pentano ≥99% (by GC), CHROMASOLV™ per analisi dei pesticidi, Riedel-de Haën™
Anbieter: Honeywell Chemicals
Acetone, extra dry 99.8%, AcroSeal®
Anbieter: Thermo Fisher Scientific
2-Propanolo, BAKER, Laboratory Reagent, J.T.Baker®
Anbieter: Avantor
Ciclopentano 95+%, puro
Anbieter: Thermo Fisher Scientific
Xilene (miscela di isomeri) ≥98%, liquido
Anbieter: PanReac AppliChem
Acetone ≥99.5% ACS
Anbieter: VWR Chemicals