cis-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)cyclopropanecarboxylic acid 1 * 1 g
Catalog # PHBLPBWZ135-1G
Supplier: Pharmablock (USA)
cis-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)cyclopropanecarboxylic acid 1 * 1 g
Catalog # PHBLPBWZ135-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Acqua, AnalaR NORMAPUR® ISO 3696 Grade 3 per analisi
Supplier: VWR Chemicals
Glicerina 85% Ph. Eur.
Supplier: VWR Chemicals
Acqua, LiChrosolv® tested for UHPLC-MS, Supelco®
Supplier: Merck
n-Esano, LiChrosolv® for liquid chromatography, Supelco®
Supplier: Merck
Formammide ≥99.0%, AnalaR NORMAPUR® per analisi
Supplier: VWR Chemicals
Glicerina ≥99.5%, AnalaR NORMAPUR® ACS per analisi, bidistillata
Supplier: VWR Chemicals
Acqua, HiPerSolv CHROMANORM® per HPLC
Supplier: VWR Chemicals
Toluene ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS per analisi
Supplier: VWR Chemicals
Metanolo ≥99.9% (by GC), HiPerSolv CHROMANORM® per LC-MS, adatto per UPLC/UHPLC
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
n-Esano ≥95%, TECHNICAL
Supplier: VWR Chemicals
Acqua, HiPerSolv CHROMANORM® per LC-MS, adatto per UPLC/UHPLC
Supplier: VWR Chemicals
Etanolo assoluto ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS per analisi
Supplier: VWR Chemicals
Etanolo 96% (v/v), EMPROVE® EXPERT Ph. Eur., BP, ChP, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS