spiro[5,7-dihydrocyclopenta[b]pyridine-6,4'-piperidine]-5-amine trihydrochloride 1 * 1 g
Catalog # PHBLPBV1168-1-1G
Supplier: Pharmablock (USA)
spiro[5,7-dihydrocyclopenta[b]pyridine-6,4'-piperidine]-5-amine trihydrochloride 1 * 1 g
Catalog # PHBLPBV1168-1-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Etere etilico ≥99% stabilizzato, GPR RECTAPUR®
Supplier: VWR Chemicals
ter-Butil metil etere, LiChrosolv® per HPLC, Supelco®
Supplier: Merck
Etanolo 70% ≥70% (v/v), TechniSolv®
Supplier: VWR Chemicals
Etanolo, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Etilen glicole ≥99.7%, AnalaR NORMAPUR® per analisi
Supplier: VWR Chemicals
2,2,4-Trimetil pentano ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS per analisi
Supplier: VWR Chemicals
Tetraidrofurano ≥99% stabilizzato, GPR RECTAPUR®
Supplier: VWR Chemicals
Acqua, AnalaR NORMAPUR® ISO 3696 Grade 3 per analisi
Supplier: VWR Chemicals
Toluene ≥98%, TECHNICAL
Supplier: VWR Chemicals
Etilen glicole ≥98%, TECHNICAL
Supplier: VWR Chemicals
Cicloesano ≥99.5%, PESTINORM® per analisi dei pesticidi
Supplier: VWR Chemicals
Piridina ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS per analisi
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS per analisi
Supplier: VWR Chemicals
Diclorometano ≥99% stabilizzato, GPR RECTAPUR®
Supplier: VWR Chemicals
Metanolo ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS per analisi
Supplier: VWR Chemicals