O1-tert-butyl O2-methyl (2R)-2,3-dihydropyrrole-1,2-dicarboxylate 1 * 1 g
Catalog # PHBLPBV1087-1G
Supplier: Pharmablock (USA)
O1-tert-butyl O2-methyl (2R)-2,3-dihydropyrrole-1,2-dicarboxylate 1 * 1 g
Catalog # PHBLPBV1087-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Metanolo, anidro (max. 0,0020% H<sub>2</sub>O) ≥99.9%
Supplier: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® per HPLC
Supplier: VWR Chemicals
Etile acetato ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
1-Butanolo ≥98.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
ter-Butil metil etere, EMSURE® ACS per analisi, Supelco®
Supplier: Merck
2,2,4-Trimetil pentano, anidro (max. 0,01% H<sub>2</sub>O) ≥99%, GPR RECTAPUR® per sintesi
Supplier: VWR Chemicals
(±)-2-Butanolo, EMSURE® per analisi, Supelco®
Supplier: Merck
Metanolo ≥99.9%, HiPerSolv CHROMANORM®, PFAS grade per LC-MS
Supplier: VWR Chemicals
Acetonitrile, ChromAR® per HPLC, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
n-Eptano
Supplier: Merck
N-Metil-2-pirrolidone (NMP) 99+% ACS
Supplier: Thermo Fisher Scientific
2,2,4-Trimetil pentano ≥99% (by GC), CHROMASOLV™ per analisi dei pesticidi, Riedel-de Haën™
Supplier: Honeywell Chemicals
n-Eptano 95%
Supplier: Thermo Fisher Scientific
Cicloesano 99+% ACS, meets analytical specifications of Reagent Ph. Eur. per spettroscopia
Supplier: Thermo Fisher Scientific
2-Etossietanolo ≥99%
Supplier: Thermo Fisher Scientific