4-bromo-N,N-dimethyl-3-(trifluoromethyl)pyrazole-1-sulfonamide 1 * 1 g
Catalog # PHBLPBU1936-1G
Supplier: Pharmablock (USA)
4-bromo-N,N-dimethyl-3-(trifluoromethyl)pyrazole-1-sulfonamide 1 * 1 g
Catalog # PHBLPBU1936-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
ACETONITRILE EMPLURA® 1 * 4 L
Supplier: Merck
Acqua, GPR RECTAPUR®
Supplier: VWR Chemicals
Diclorometano ≥99% stabilizzato, GPR RECTAPUR®
Supplier: VWR Chemicals
Etilen glicole 99.5% per analisi
Supplier: Thermo Fisher Scientific
Cloroformio 99.0-99.4% (by GC) stabilizzato, Puriss. p.a. ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Cicloesano 99.8% per HPLC
Supplier: Thermo Fisher Scientific
Toluene ≥99.7% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Cesio cloruro 99.99% (trace metals basis), Optical Grade per spettroscopia
Supplier: Thermo Fisher Scientific
1-Pentanolo 99+% ACS
Supplier: Thermo Fisher Scientific
DIMETHYL SULFOXIDE (USP, BP, PH. EUR.) 1 * 190 L
Supplier: PanReac AppliChem
ISOAMYL ALCOHOL ACCORDING TO GERBER FOR 1 * 1.000 mL
Supplier: PanReac AppliChem
n-Pentano 99+%, extra puro
Supplier: Thermo Fisher Scientific
Diclorometano ≥99.9% (by GC) stabilizzato, Puriss. p.a. ACS, ISO analytical reagent, Riedel-de Haën™
Supplier: Honeywell Chemicals
2-Propanolo ≥99.8% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals