5-(4-methoxy-1-piperidyl)pyridin-2-amine 1 * 1 g
PHBLPBU0330-1G
Frequently Bought Together
Etanolo, GPR RECTAPUR®
: VWR Chemicals
Acqua, HiPerSolv CHROMANORM® per LC-MS, adatto per UPLC/UHPLC
: VWR Chemicals
Etanolo 96% vol Ph. Eur.
: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Acqua, GPR RECTAPUR®
: VWR Chemicals
Glicerina ≥99.5%, AnalaR NORMAPUR® ACS per analisi, bidistillata
: VWR Chemicals
Acqua, HiPerSolv CHROMANORM® per HPLC
: VWR Chemicals
Etanolo 96% (v/v), AnalaR NORMAPUR® per analisi
: VWR Chemicals
n-Eptano ≥99.0%, AnalaR NORMAPUR® per analisi
: VWR Chemicals
2-Propanolo ≥98%, TECHNICAL
: VWR Chemicals