2-chloro-3-methoxypyrazine 1 * 5 g
Catalog # PHBLPBTEN19128-5G
Supplier: Pharmablock (USA)
Frequently Bought Together
Etanolo 70% ≥70% (v/v), TechniSolv®
Supplier: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® per HPLC
Supplier: VWR Chemicals
Etanolo, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® per LC-MS, adatto per UPLC/UHPLC
Supplier: VWR Chemicals
Acqua, HiPerSolv CHROMANORM® per LC-MS, adatto per UPLC/UHPLC
Supplier: VWR Chemicals
Etanolo assoluto ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS per analisi
Supplier: VWR Chemicals
Metanolo ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., grado gradiente per HPLC
Supplier: VWR Chemicals
Acetonitrile, LiChrosolv®, hypergrade per LC-MS, Supelco®
Supplier: Merck
Acqua, HiPerSolv CHROMANORM® per HPLC
Supplier: VWR Chemicals
Acqua, LiChrosolv® per cromatografia, per LC-MS, Supelco®
Supplier: Merck
2-Propanolo ≥99.8%, HiPerSolv CHROMANORM® per HPLC
Supplier: VWR Chemicals