imidazo[1,2-a]pyrazine 1 * 5 g
Catalog # PHBLPBTEN19003-5G
Supplier: Pharmablock (USA)
Frequently Bought Together
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® per LC-MS, adatto per UPLC/UHPLC
Supplier: VWR Chemicals
Acetonitrile, LiChrosolv®, hypergrade per LC-MS, Supelco®
Supplier: Merck
Acqua, HiPerSolv CHROMANORM® per HPLC
Supplier: VWR Chemicals
Metanolo ≥99.9% (by GC), HiPerSolv CHROMANORM® per LC-MS, adatto per UPLC/UHPLC
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Glicerina 85% Ph. Eur.
Supplier: VWR Chemicals
Diclorometano ≥99.5% stabilizzato, AnalaR NORMAPUR® Reag. Ph. Eur., ACS per analisi
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS per analisi
Supplier: VWR Chemicals
2-Propanolo ≥98%, TECHNICAL
Supplier: VWR Chemicals
Etere etilico ≥99% stabilizzato, GPR RECTAPUR®
Supplier: VWR Chemicals
Xilene (miscela di isomeri) ≥98%, TECHNICAL
Supplier: VWR Chemicals
Acqua, LiChrosolv® per cromatografia, per LC-MS, Supelco®
Supplier: Merck
Etanolo assoluto ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS per analisi
Supplier: VWR Chemicals
Acqua, HiPerSolv CHROMANORM® per LC-MS, adatto per UPLC/UHPLC
Supplier: VWR Chemicals
Etanolo 70% ≥70% (v/v), TechniSolv®
Supplier: VWR Chemicals