2-pyrrolidin-1-yl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-3-carbonitrile 1 * 1 g
Catalog # PHBLPBTD135-1G
Supplier: Pharmablock (USA)
2-pyrrolidin-1-yl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-3-carbonitrile 1 * 1 g
Catalog # PHBLPBTD135-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
p-Xilene 99% per spettroscopia
Supplier: Thermo Fisher Scientific
N-Metil-2-pirrolidone (NMP) 99.5% per analisi
Supplier: Thermo Fisher Scientific
n-Esano 95+%, extra puro
Supplier: Thermo Fisher Scientific
(±)-2-Metiltetraidrofurano 99+% stabilizzato, puro
Supplier: Thermo Fisher Scientific
2,2,4-Trimetil pentano 99.5% per analisi
Supplier: Thermo Fisher Scientific
2-Propanolo ≥99.8% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
TRICHLOROACETIC ACID (BP, PH. EUR.) PURE 1 * 5 kg
Supplier: PanReac AppliChem
Etanolammina ACS
Supplier: Thermo Fisher Scientific
n-Esano
Supplier: Roth Carl
Acetone ≥99.5% ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Diisopropile ossido ≥98.5% (by GC) stabilizzato, Puriss. p.a., Riedel-de Haën™
Supplier: Honeywell Chemicals
Anisolo, extra dry 99%, AcroSeal®
Supplier: Thermo Fisher Scientific
Glicerina ≥83.5-88.5% (w/w) in soluzione acquosa, puro
Supplier: Thermo Fisher Scientific