methyl 4-bromo-2-formyl-benzoate 1 * 1 g
Catalog # PHBLPBT8024-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Acetonitrile ≥99.95%, HiPerSolv CHROMANORM® ULTRA per LC-MS, per UPLC/UHPLC-MS
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Metanolo ≥98.5%, TECHNICAL
Supplier: VWR Chemicals
Metanolo ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., grado gradiente per HPLC
Supplier: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® per HPLC
Supplier: VWR Chemicals
Etanolo 96% (v/v), AnalaR NORMAPUR® per analisi
Supplier: VWR Chemicals
n-Esano ≥95%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS per analisi
Supplier: VWR Chemicals
Etere di petrolio, 40…65 °C, anidro, TECHNICAL (max. 0,0100% idrocarburi aromatici)
Supplier: VWR Chemicals
Metanolo ≥99.8%, HiPerSolv CHROMANORM®, grado isocratico per HPLC
Supplier: VWR Chemicals
Acqua, HiPerSolv CHROMANORM® per LC-MS, adatto per UPLC/UHPLC
Supplier: VWR Chemicals
Etanolo assoluto ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS per analisi
Supplier: VWR Chemicals
Metanolo ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS per analisi
Supplier: VWR Chemicals