(3aR,4R,6R,6aR)-4-methoxy-2,2,6-trimethyl-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3]dioxole 1 * 1 g
PHBLPBT2929-1G
(3aR,4R,6R,6aR)-4-methoxy-2,2,6-trimethyl-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3]dioxole 1 * 1 g
PHBLPBT2929-1G
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Etanolo 96% (v/v), AnalaR NORMAPUR® per analisi
: VWR Chemicals
2-Propanolo ≥99.8%, HiPerSolv CHROMANORM® per HPLC
: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS per analisi
: VWR Chemicals
Cloroformio ≥99.8% stabilizzato, HiPerSolv CHROMANORM® per HPLC
: VWR Chemicals