tert-butyl (1S,5R)-6-oxo-3-azabicyclo[3.2.0]heptane-3-carboxylate 1 * 1 g
PHBLPBLJD0882-1G
tert-butyl (1S,5R)-6-oxo-3-azabicyclo[3.2.0]heptane-3-carboxylate 1 * 1 g
PHBLPBLJD0882-1G
Frequently Bought Together
Etanolo, GPR RECTAPUR®
: VWR Chemicals
Etanolo 70% ≥70% (v/v), TechniSolv®
: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Etanolo 96% vol Ph. Eur.
: VWR Chemicals
Glicerina ≥99.5%, AnalaR NORMAPUR® ACS per analisi, bidistillata
: VWR Chemicals
Acqua, TECHNICAL, demineralizzata
: VWR Chemicals
Diclorometano ≥99% stabilizzato, GPR RECTAPUR®
: VWR Chemicals
Etere etilico ≥99% stabilizzato, GPR RECTAPUR®
: VWR Chemicals
Etile acetato ≥99.7%, PESTINORM® per analisi dei pesticidi
: VWR Chemicals