(2S,4S,5S)-1-tert-butoxycarbonyl-4-hydroxy-5-methyl-pyrrolidine-2-carboxylic acid 1 * 1 g
Catalog # PHBLPBLJD0540-1G
Supplier: Pharmablock (USA)
(2S,4S,5S)-1-tert-butoxycarbonyl-4-hydroxy-5-methyl-pyrrolidine-2-carboxylic acid 1 * 1 g
Catalog # PHBLPBLJD0540-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Etere di petrolio, 60…80 °C, AnalaR NORMAPUR® per analisi
Supplier: VWR Chemicals
ter-Butil metil etere, EMSURE® ACS per analisi, Supelco®
Supplier: Merck
n-Eptano, EMPLURA®, Supelco®
Supplier: Merck
2-Propanolo, EMPROVE® ESSENTIAL Ph. Eur., BP, JP, USP, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Acqua, extra puro, deionised
Supplier: Thermo Fisher Scientific
Etilen glicole 99.5% per analisi
Supplier: Thermo Fisher Scientific
Tetraidrofurano, anidro 99.9% stabilizzato, extra puro
Supplier: Thermo Fisher Scientific
p-Xilene 99%, puro
Supplier: Thermo Fisher Scientific
Acetonitrile ≥99.9%, CHROMASOLV™, grado gradiente per HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
ter-Butanolo ≥99%
Supplier: Thermo Fisher Scientific
Cicloesano ≥99.5% (by GC), CHROMASOLV™ per analisi dei pesticidi, Riedel-de Haën™
Supplier: Honeywell Chemicals
Acetonitrile 99+% per spettroscopia
Supplier: Thermo Fisher Scientific
Metanolo 99.9% per spettroscopia
Supplier: Thermo Fisher Scientific