cis-2-aminocyclohexanecarboxylic acid hydrochloride 1 * 1 g
PHBLPBJLD0003-1-1G
Frequently Bought Together
2-Propanolo ≥98%, TECHNICAL
: VWR Chemicals
Etanolo assoluto, EMPROVE® EXPERT Ph. Eur., BP, ChP, JP, USP adatto a essere utilizzato come eccipiente, SAFC®
: MERCK PRODUCTION CHEMICALS
FAM Benzine DIN 51635, AnalaR NORMAPUR® per analisi
: VWR Chemicals
2-Propanolo, extra dry 99.8%, AcroSeal®
: Thermo Fisher Scientific
N-Metil-2-pirrolidone (NMP) 99+% ACS
: Thermo Fisher Scientific
Etilen glicole 99.5% per analisi
: Thermo Fisher Scientific
1-Butanolo 99.5% ACS, meets analytical specifications of Reagent Ph. Eur.
: Thermo Fisher Scientific
Dimetilsolfossido ≥99.7%, CHROMASOLV™ Plus per HPLC, Riedel-de Haën™
: Honeywell Chemicals
MORPHISTO METHYLENBLAU 1 %, WÄSSRIG 1 * 2.500 ml
: MARKET SOURCE PART PROCESS
n-Eptano 95%
: Thermo Fisher Scientific