2-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3-benzothiazole 1 * 1 g
PHBLPBJL8005-1G
2-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3-benzothiazole 1 * 1 g
PHBLPBJL8005-1G
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Acqua, HiPerSolv CHROMANORM® per HPLC
: VWR Chemicals
n-Eptano ≥99%, GPR RECTAPUR®
: VWR Chemicals
Diclorometano ≥98% stabilizzato, TECHNICAL
: VWR Chemicals
Clorobenzene ≥99%, GPR RECTAPUR®
: VWR Chemicals
Etanolo 70% ≥70% (v/v), TechniSolv®
: VWR Chemicals
n-Esano ≥95%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS per analisi
: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS per analisi
: VWR Chemicals
2-Propanolo ≥98%, TECHNICAL
: VWR Chemicals