7-bromo-4H-pyrido[1,2-a]pyrimidin-4-one 1 * 1 g
Catalog # PHBLPB95773-1G
Supplier: Pharmablock (USA)
7-bromo-4H-pyrido[1,2-a]pyrimidin-4-one 1 * 1 g
Catalog # PHBLPB95773-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
2-Propanolo ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS per analisi
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Metanolo ≥99.9% (by GC), HiPerSolv CHROMANORM® per LC-MS, adatto per UPLC/UHPLC
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® per LC-MS, adatto per UPLC/UHPLC
Supplier: VWR Chemicals
Etanolo assoluto ≥99.5%, EMPLURA®, Supelco®
Supplier: Merck
Glicerina ≥99.5%, AnalaR NORMAPUR® ACS per analisi, bidistillata
Supplier: VWR Chemicals
Toluene ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS per analisi
Supplier: VWR Chemicals
Metanolo ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., grado gradiente per HPLC
Supplier: VWR Chemicals
Acqua, TECHNICAL, demineralizzata
Supplier: VWR Chemicals
2-Propanolo 70% ≥70% (v/v), GPR RECTAPUR®
Supplier: VWR Chemicals
Etere di petrolio, 40…60 °C, GPR RECTAPUR®
Supplier: VWR Chemicals
Etanolo assoluto ≥99.8%, HiPerSolv CHROMANORM®, grado gradiente per HPLC
Supplier: VWR Chemicals
Glicerolo (glicerina) USP, Ph. Eur., BP
Supplier: VWR Chemicals