methyl 2-chloro-6-methylpyridine-4-carboxylate 1 * 10 g
Catalog # PHBLPB05828-10G
Supplier: Pharmablock (USA)
methyl 2-chloro-6-methylpyridine-4-carboxylate 1 * 10 g
Catalog # PHBLPB05828-10G
Supplier: Pharmablock (USA)
Frequently Bought Together
Acqua, LiChrosolv® per cromatografia, per LC-MS, Supelco®
Supplier: Merck
Metanolo ≥99.8%, HiPerSolv CHROMANORM®, grado isocratico per HPLC
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Metanolo ≥99.9% (by GC), HiPerSolv CHROMANORM® per LC-MS, adatto per UPLC/UHPLC
Supplier: VWR Chemicals
Metanolo ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., grado gradiente per HPLC
Supplier: VWR Chemicals
2-Propanolo ≥99.8%, HiPerSolv CHROMANORM® per HPLC
Supplier: VWR Chemicals
Etanolo, GPR RECTAPUR®
Supplier: VWR Chemicals
Etanolo 70% ≥70% (v/v), TechniSolv®
Supplier: VWR Chemicals
Acqua, HiPerSolv CHROMANORM® per LC-MS, adatto per UPLC/UHPLC
Supplier: VWR Chemicals
Etanolo assoluto ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS per analisi
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® per LC-MS, adatto per UPLC/UHPLC
Supplier: VWR Chemicals
n-Esano ≥95%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS per analisi
Supplier: VWR Chemicals