1-methylcyclopropane-1-carboxylic acid 1 * 5 g
Catalog # PHBLPB05812-5G
Supplier: Pharmablock (USA)
Frequently Bought Together
n-Pentano ≥99.8%, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
Diclorometano ≥99.5% stabilizzato, AnalaR NORMAPUR® Reag. Ph. Eur., ACS per analisi
Supplier: VWR Chemicals
Toluene : 2-Propanol : Water (49,5 : 50 : 0,5) % vol., VOLUSOL® titration solvent
Supplier: VWR Chemicals
(±)-2-Metiltetraidrofurano ≥99.0% stabilizzato, GPR RECTAPUR® per sintesi
Supplier: VWR Chemicals
Etanolammina, EMSURE® per analisi, Supelco®
Supplier: Merck
n-Esano, LiChrosolv® for liquid chromatography, Supelco®
Supplier: Merck
Esano (miscela di isomeri), TECHNICAL
Supplier: VWR Chemicals
Metanolo, Prepsolv® for preparative chromatography, Supelco®
Supplier: Merck
Etanolo Eurodenatured ≥70%, TechniSolv® (1:1:1)
Supplier: VWR Chemicals
2-Propanolo ≥99.8%, HiPerSolv CHROMANORM® per HPLC
Supplier: VWR Chemicals
Polivinil alcool, M.W. ∼ 13,000 98%, hydrolysed
Supplier: Thermo Fisher Scientific
Alcol benzilico 99-100.5% (by GC), Puriss., meets analytical specification of BP, Ph. Eur., NF, Riedel-de Haën™
Supplier: Honeywell Chemicals
GLYCEROL (USP, BP, PH. EUR.) PURE, PHARM 1 * 1.000 mL
Supplier: PanReac AppliChem