Plastic feeding tubes, 16ga x 38mm. 10 sterile pouches of 5. Polyurethane tube with rounded tip. For mouse oral gavage. Disposable. Designed to minimize trauma to esophagus. Large ID for viscous compounds. Straight tube wipes clean to prevent aspirat 1 * 50 ST
Codice catalogo MSPPFTPU-16-38-50
Nome del fornitore: MARKET SOURCE PART PROCESS
Frequently Bought Together