Terminal disinfectant for open plant application, QAC based 1 * 1 Bottle
Catalog # DIVY101108286
Supplier: Diversey
Terminal disinfectant for open plant application, QAC based 1 * 1 Bottle
Catalog # DIVY101108286
Supplier: Diversey
Frequently Bought Together
Etilen glicole ≥99.7%, AnalaR NORMAPUR® per analisi
Supplier: VWR Chemicals
Xilene (miscela di isomeri) ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS per analisi
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, grado gradiente per HPLC
Supplier: VWR Chemicals
Acqua, AnalaR NORMAPUR® ISO 3696 Grade 3 per analisi
Supplier: VWR Chemicals
n-Esano, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
Etanolo 96% (v/v), AnalaR NORMAPUR® per analisi
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® per LC-MS, adatto per UPLC/UHPLC
Supplier: VWR Chemicals
Acetonitrile, LiChrosolv®, hypergrade per LC-MS, Supelco®
Supplier: Merck
Toluene ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS per analisi
Supplier: VWR Chemicals
Diclorometano ≥98% stabilizzato, TECHNICAL
Supplier: VWR Chemicals
2-Propanolo ≥98%, TECHNICAL
Supplier: VWR Chemicals
2-Propanolo ≥99.8%, HiPerSolv CHROMANORM® per HPLC
Supplier: VWR Chemicals
Xilene (miscela di isomeri) ≥98%, TECHNICAL
Supplier: VWR Chemicals
Etanolo assoluto, EMPROVE® EXPERT Ph. Eur., BP, ChP, JP, USP adatto a essere utilizzato come eccipiente, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Etanolo assoluto ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS per analisi
Supplier: VWR Chemicals