Masterflex® Fitting, Polycarbonate, Straight, Flexible PVC Tubing with Male/Male Luer Ends, 24" Long; 10/PK
Codice catalogo MFLX30600-62
Nome del fornitore: Avantor Fluid Handling
Informazioni su questo articolo
Get a seamless transition from source to system.
- Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Connection Type: male luer lock x male luer lock with 24" long tubing
This high pressure flexible PVC tubing has luer fittings at each end and comes in various lengths. Can be sterilised by ETO.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
- Tipo di connessione:Tubing Assembly
- Flow pattern:Straight
- Descrizione:Fitting, Polycarbonate, Straight, Flexible PVC Tubing with Male/Male Luer Ends, 24" Long; 10/PK
- Seq:06
- Environmentally Preferable:
- Modello:30600-62
- Connection type (port 1):Male luer lock
- Connection type (port 2):Male luer lock
- No. of ports:2
- Garanzia:Out of box
- Conf:10
- Fitting material:Polycarbonate
Frequently Bought Together