STERI-WATER is a quality water which is designed for operations that require a USP grade water for chemical formulations, cleaning, and rinsing. Not for human or animal injection, diagnostic, or therapeutic use. 1 * 2 Bottle
VELTSTWA-2G
STERI-WATER is a quality water which is designed for operations that require a USP grade water for chemical formulations, cleaning, and rinsing. Not for human or animal injection, diagnostic, or therapeutic use. 1 * 2 Bottle
VELTSTWA-2G
Frequently Bought Together
Etere etilico ≥99% stabilizzato, GPR RECTAPUR®
: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Acqua, HiPerSolv CHROMANORM® per LC-MS, adatto per UPLC/UHPLC
: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS per analisi
: VWR Chemicals
n-Esano ≥95%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS per analisi
: VWR Chemicals