Process2Clean 5 is a highly built neutral pH cleaning agent that can be used as a stand-alone detergent or as an additive to improve the cleaning performance of all VAI cleaning agents. Ideal for use on most all substrate material and effective on m 1 * 1 Bottle
Codice catalogo VELTPC-5-5G-01
Nome del fornitore: VELTEK ASSOCIATES
Frequently Bought Together