The DEC-50-FR Foot Pedal is an optional accessory that attaches to the DEC-50 and allows the user to press a Foot Pedal to dispense select VAI Aerosol products. 1 * 1 items
Codice catalogo VELTDEC-50-FR
Nome del fornitore: VELTEK ASSOCIATES
Frequently Bought Together