25l stainless steel drum with 1 opening incl. screw cap with pressure regulating valve at the top, with additional 1 clamp at the top incl. blind cap, gasket and fixing bracket, with level indicator on the side 1 * 1 pezzi
Codice catalogo ROETSA_25LICLAMP
Nome del fornitore: ROETZMEIER SICHERH BEHAELTER
Frequently Bought Together