DEC-CLEAN SIMPLEMIX RESIDUE REMOVER 3.79 LITER 1 * 4 items
Katalognr. VELTDC-05-1Z-E
Anbieter: VELTEK ASSOCIATES
DEC-CLEAN SIMPLEMIX RESIDUE REMOVER 3.79 LITER 1 * 4 items
Katalognr. VELTDC-05-1Z-E
Anbieter: VELTEK ASSOCIATES
Frequently Bought Together
Metanolo ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS per analisi
Anbieter: VWR Chemicals
Acqua USP, JP, Ph. Eur., purificato
Anbieter: VWR Chemicals
Safety Bottle Container
Anbieter: Merck
Acetone ≥99%, TECHNICAL
Anbieter: VWR Chemicals
Acetonitrile ≥99.95%, HiPerSolv CHROMANORM® ULTRA per LC-MS, per UPLC/UHPLC-MS
Anbieter: VWR Chemicals
Acetonitrile, LiChrosolv®, hypergrade per LC-MS, Supelco®
Anbieter: Merck
Etere etilico ≥99.5% stabilizzato, HiPerSolv CHROMANORM® per HPLC
Anbieter: VWR Chemicals
Acido trifluoroacetico, Sigma-Aldrich®
Anbieter: Merck
Acetonitrile ≥99%, TECHNICAL
Anbieter: VWR Chemicals
2-Etossietanolo ≥99%, TECHNICAL
Anbieter: VWR Chemicals
Diclorometano ≥99% stabilizzato, GPR RECTAPUR®
Anbieter: VWR Chemicals
Piridina ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS per analisi
Anbieter: VWR Chemicals