[EN]65ML PL POLYACRYLATE 2 (65 TESTS) 1 * 65 mL
WTIDPL65PLYA2
Frequently Bought Together
Formaldeide 4% stabilizzato, buffered (pH 7,0 ±0,1)
: VWR Chemicals
UltraClear® per istologia, J.T.Baker®
: Avantor
Formalin 10%, Q Path®, buffered
: VWR Chemicals
NANOCOLOR® Photometric Tube Test Kits for Nitrite
: MACHEREY-NAGEL
SODIUM HYDROXIDE SOLUTION 6 MOL/L EMPROV 1 * 1 L
: MERCK PRODUCTION CHEMICALS
Strisce reattive, perossidi, Dosatest
: VWR Chemicals
Inchiostro, verde, Q Path® stain for histology
: VWR Chemicals
ADF Volumetric Solution for analysis of animal feed (NF V18-122)
: VWR Chemicals
VWR® Q Path® O.C.T. Compound Mounting Medium for Cryotomy
: VWR Chemicals
BIOSYSTEMS PHOSPHATE BUFFERED SALINE PLUS TWEEN 20 (20X) PH 7.
: MARKET SOURCE PART PROCESS