[EN]REAGENT KIT FOR PRIMELAB ID47 (65 TE 1 * 1 pezzi
WTIDPL47-KIT
Frequently Bought Together
Safranina O, Certifiable Grade
: VWR Chemicals
INSTANT EOSIN ALOCHOLIC- 1 * 1 pezzi
: Epredia
Fast Red RC Salt, Sigma-Aldrich®
: SIGMA ALDRICH MICROSCOPY
[EN]19 COMPOSANTS DANS HNO3 5% 1 * 100 mL
: CUSTOM MADE CHEMICALS LAB
[EN]60G PL SILICA HR 2 (241 TESTS) 1 * 60 g
: WATER ID PRIMELAB
Sodio idrossido 30% in water, grado tecnico
: Thermo Fisher Scientific
Solvent black 5, Sigma-Aldrich®
: SIGMA ALDRICH MICROSCOPY
Ammoniaca soluzione 28,0 - 30,0% ACS
: Thermo Fisher Scientific