[EN]REAGENT KIT FOR PRIMELAB ID159(285TE 1 * 1 pezzi
WTIDPL159-KIT
Frequently Bought Together
Compresse Kjeldahl
: VWR Chemicals
Acido trifluoroacetico ≥99.9%, HiPerSolv CHROMANORM® per LC-MS
: VWR Chemicals
Sodio idrossido 97.0-100.5%, in pastiglie NF, JP, Ph. Eur., BP
: VWR Chemicals
Trypan blue 0,4% in soluzione acquosa, Sigma-Aldrich®
: SIGMA ALDRICH MICROSCOPY
Anthrone in ACS Reagent grade water 97%, Sigma-Aldrich®
: SIGMA ALDRICH MICROSCOPY
HYDROCHLORIC ACID 0,01 N IN PROPANOL-2 1 * 1 L
: PROLABO CMP