[EN]REAGENT KIT FOR PRIMELAB ID151(50 TE 1 * 1 pezzi
Catalog # WTIDPL151-KIT
Supplier: WATER ID PRIMELAB
[EN]REAGENT KIT FOR PRIMELAB ID151(50 TE 1 * 1 pezzi
Catalog # WTIDPL151-KIT
Supplier: WATER ID PRIMELAB
Frequently Bought Together
Acido formico ≥99%, HiPerSolv CHROMANORM® per LC-MS
Supplier: VWR Chemicals
Kit analitici, test Ammonio per Spectroquant®, Supelco®
Supplier: Merck
Kit analitici, COD in cuvetta per Spectroquant®, Supelco®
Supplier: Merck
Kit analitici, tensioattivi, per Spectroquant®, Supelco®
Supplier: Merck
Immersion oil for microscopy, Sigma-Aldrich®
Supplier: Merck
Kit analitici, test fosfato per Spectroquant®, Supelco®
Supplier: Merck
Kit analitici, nitrati per Spectroquant®, Supelco®
Supplier: Merck
BIOSYSTEMS Bond Epitope Retrieval Lösung 2 CE/IVD RT
Supplier: MARKET SOURCE PART PROCESS
Reagents for Phosphate Determination using Lovibond® MD photometers
Supplier: Lovibond
Test kit, fotometrico, Zinco, test in cuvetta LCK
Supplier: Hach
Acid Red 183 30% (dye content), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Powder pillows, ammonia
Supplier: Hach
Nitrite VHR L, 500 ml
Supplier: Lovibond