METHYL 1-METHYL-5-(4,4,5,5-TETRAMETHYL-1,3,2-DIOXABOROLAN-2-YL)PYRAZOLE-3-CARBOXYLAT
Catalog # PHBLPBXAA892-1G
Supplier: Pharmablock (USA)
METHYL 1-METHYL-5-(4,4,5,5-TETRAMETHYL-1,3,2-DIOXABOROLAN-2-YL)PYRAZOLE-3-CARBOXYLAT
Catalog # PHBLPBXAA892-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
2-Propanolo ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS per analisi
Supplier: VWR Chemicals
Etile acetato ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® per LC-MS, adatto per UPLC/UHPLC
Supplier: VWR Chemicals
Etanolo 96% (v/v), AnalaR NORMAPUR® per analisi
Supplier: VWR Chemicals
2-Propanolo, SupraSolv® for gas chromatography, Supelco®
Supplier: Merck
Metanolo ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., grado gradiente per HPLC
Supplier: VWR Chemicals
Etanolo 96% vol Ph. Eur.
Supplier: VWR Chemicals
Metanolo, SupraSolv® for gas chromatography, Supelco®
Supplier: Merck
Acqua, HiPerSolv CHROMANORM® per LC-MS, adatto per UPLC/UHPLC
Supplier: VWR Chemicals
Etanolo assoluto ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS per analisi
Supplier: VWR Chemicals
Acqua, LiChrosolv® per cromatografia, per LC-MS, Supelco®
Supplier: Merck