Digital Solutions
Applications
Support
About
Inserisci ordini
Stato dell'ordine
Il tuo profilo
Carrello
Switzerland
Inserisci ordini
Stato dell'ordine
Il tuo profilo
Carrello
ContactUsLinkComponent
[EN]5 COMPONENTS IN WATER 1 * 500 mL
CMPLRM092682L5
:
CUSTOM MADE CHEMICALS LAB
[EN]5 COMPONENTS IN WATER 1 * 500 mL
CMPLRM092682L5
:
CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
Compresse Kjeldahl
: VWR Chemicals
Dihydrorhodamine 123 ≥95% (by HPLC), Sigma-Aldrich®
: SIGMA ALDRICH MICROSCOPY
Sodio idrossido, anidro ≥98%, in pastiglie, per analisi
: Honeywell Chemicals
[EN]BIOTECTOR 1.8N SULFURIC ACID 80MG/L 1 * 20 L
: Hach
Reagents for Phosphate Determination using Lovibond® MD photometers
: Lovibond
Diluid 610 per ematologia, J.T.Baker®
: Avantor
Reagents for determination of ammonia and total kjeldahl nitrogen by the nessler method
: Hach