[EN]INOSITOL NF 1 * 500 g
Catalog # SPCMI1012-500GM
Supplier: Spectrum Chemical
Frequently Bought Together
Glicerina ≥98%, GPR RECTAPUR®
Supplier: VWR Chemicals
Etere etilico stabilizzato, EMPLURA®, Supelco®
Supplier: Merck
Anilina per sintesi, Sigma-Aldrich®
Supplier: Merck
2-Propanolo ≥70% (v/v), SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Xilene (miscela di isomeri), extra dry 98+%, AcroSeal®
Supplier: Thermo Fisher Scientific
n-Esano ≥95% per spettrofotometria
Supplier: Thermo Fisher Scientific
Acetone ≥99.8%, CHROMASOLV™ per HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
N-(1-Naftil) etilendiammina dicloridrato, Supelco®
Supplier: Merck
Etile acetato ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
N,N-Dimetilformammide ≥99.7% per HPLC
Supplier: Thermo Fisher Scientific
Liquido per manometro
Supplier: VWR Chemicals
1,4-Diossano, extra dry over Molecular Sieve 99,5% stabilizzato, AcroSeal®
Supplier: Thermo Fisher Scientific