[EN]CUSTOM MADE STANDARD 20 COMP 1 * 500 mL
Catalog # CMPLRM091165L5
Supplier: CUSTOM MADE CHEMICALS LAB
[EN]CUSTOM MADE STANDARD 20 COMP 1 * 500 mL
Catalog # CMPLRM091165L5
Supplier: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
Kit analitici, COD in cuvetta per Spectroquant®, Supelco®
Supplier: Merck
Eosina Y in soluzione acquosa, Q Path® per microscopia
Supplier: VWR Chemicals
Papanicolaou's solution 2a Orange G (OG 6), Sigma-Aldrich®
Supplier: Merck
UltraClear® per istologia, J.T.Baker®
Supplier: Avantor
BUFFER pH 2 ACC. TO NF T 90-431 1 * 1 L
Supplier: PROLABO CMP
Compresse Kjeldahl, Kjeltabs
Supplier: THOMPSON & CAPPER
METHYLENE BLUE RAL 555 AQUEOUS SOLUTION 1 * 1 L
Supplier: RAL
NANOCOLOR® Photometric Tube Tests, Chloride
Supplier: MACHEREY-NAGEL
Standard per 2100AN e 2100AN IS
Supplier: Hach
Avantor® Seradigm South America Origin Fetal Bovine Serum (FBS)
Supplier: VWR Seradigm
INFIGRTINIB FREE BASE 1 * 25 mg
Supplier: ENZO LIFE SCIENCES
Nitrite VHR L, 500 ml
Supplier: Lovibond
Powder pillows, sulphate, Sulfa 4
Supplier: Lovibond
Ammoniaca soluzione 10% (w/w) in water, extra puro
Supplier: Thermo Fisher Scientific