ContactUsLinkComponent
1 component: BDE 10 [CAS:51930-04-2] 50mg/l in Iso-octane CRM, ISO 17034 and ISO 17025 Conservation 12 months. Store in a refrigerator at temperatures between 2C to 8C 1 * 1 mL
Catalog # CMPLRD0916161
1 component: BDE 10 [CAS:51930-04-2] 50mg/l in Iso-octane CRM, ISO 17034 and ISO 17025 Conservation 12 months. Store in a refrigerator at temperatures between 2C to 8C 1 * 1 mL
Catalog # CMPLRD0916161
Frequently Bought Together