5-CHLORO-6-METHOXY-1H-PYRROLO[2,3-B]PYRIDINE 1 * 10 g
PHBLPBLJ1552-10G
Frequently Bought Together
Etanolo 96% (v/v), AnalaR NORMAPUR® per analisi
: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
N,N-Dimetilformammide, extra dry over Molecular Sieve 99.8%, AcroSeal®
: Thermo Fisher Scientific
Metanolo ≥99.9%, PESTINORM® per analisi GC purge and trap
: VWR Chemicals
Etanolo, GPR RECTAPUR®
: VWR Chemicals
Metanolo, anidro (max. 0,0020% H<sub>2</sub>O) ≥99.9%
: VWR Chemicals
Metanolo ≥99.9%, CHROMASOLV™, grado gradiente per HPLC, Riedel-de Haën™
: Honeywell Chemicals
Dimetilsolfossido ≥99.7%, CHROMASOLV™ Plus per HPLC, Riedel-de Haën™
: Honeywell Chemicals