[EN]19 COMPOSANTS DANS HNO3 5% 1 * 100 mL
Catalog # CMPLRM029054L1
Supplier: CUSTOM MADE CHEMICALS LAB
[EN]19 COMPOSANTS DANS HNO3 5% 1 * 100 mL
Catalog # CMPLRM029054L1
Supplier: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
Acido trifluoroacetico ≥99.9%, HiPerSolv CHROMANORM® per LC-MS
Supplier: VWR Chemicals
Sodio idrossido 50% in soluzione acquosa, TECHNICAL
Supplier: VWR Chemicals
Potassio idrossido ≥85%, in pastiglie, GPR RECTAPUR®
Supplier: VWR Chemicals
Kit analitici, COD in cuvetta per Spectroquant®, Supelco®
Supplier: Merck
Calce sodata, AnalaR NORMAPUR® per analisi, with indicator
Supplier: VWR Chemicals
VWR®, Bustine di polvere per la determinazione del cloro, reagenti per fotometri
Supplier: VWR Chemicals
Paraffina, Punto di solidificazione: 54 - 56 °C, Q Path® per microscopia
Supplier: VWR Chemicals
Immersion oil for microscopy, Sigma-Aldrich®
Supplier: Merck
Soluzione di montaggio, Entellan® new
Supplier: Merck
Strisce reattive pH, MQuant®, Supelco®
Supplier: Merck
Kit analitici, test Ammonio per Spectroquant®, Supelco®
Supplier: Merck
Formaldeide 36% in soluzione acquosa stabilizzato, GPR RECTAPUR®
Supplier: VWR Chemicals
Allura red AC 80% (dye content), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Fast Red RC Salt, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Screw caps for glass screw top vials, 9 mm, level 1 everyday analysis, SureSTART™
Supplier: Thermo Fisher Scientific
Acid Red 183 30% (dye content), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Bromocresol green-methyl red soluzione mixed indicator, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY