phi29 Polymerase
Codice catalogo APOSBIBL0054-1000U
Nome del fornitore: Apollo Scientific
phi29 Polymerase
Codice catalogo APOSBIBL0054-1000U
Nome del fornitore: Apollo Scientific
Numero CAS:
Vai a
Product Details & Documents
Storage at −20 °C. DNA Polymerase 10 U/μl. Highly processive recombinant polymerase with exceptional strand displacement activity, which allows for highly efficient isothermal DNA amplification. The enzyme is capable of up to 70 thousands base insertions per binding event. The polymerase also possesses a 3'→5' proofreading exonuclease activity acting preferentially on ssDNA or ssRNA. Extremely high yields of amplified DNA can be obtained even from minute amounts of a template.
Specifiche
Specifiche
- Cat. No.:APOSBIBL0054-1000U
- Nome dell'enzima:phi29 Polymerase