Masterflex® Nylon Clamp for Sanitary Fittings, 1-1-1/2" Clamp Size; 10/PK
Codice catalogo MFLX00424-TO
Nome del fornitore: Avantor Fluid Handling
Informazioni su questo articolo
Collared clamp helps prevent unintentional opening.
- All fittings are compatible with Tri-Clamp® fittings
- Excellent chemical resistance; low leachable characteristics
- Descrizione:Nylon Clamp for Sanitary Fittings, 1-1-1/2" Clamp Size; 10/PK
- Materiale:Nylon
- Seq:02
- Environmentally Preferable:
- Sanitary clamp size (port 1):1-1/2"
- Sterilizzazione:Autoclave, Gamma
- Conf:10
- Tipo di morsetto:Sanitary
Frequently Bought Together