A1 BETA CASEIN (A1) BOVINE ELISA ASSAY 1 * 96 Tests
Catalog # BSENBEK-2243-1P
Supplier: Biosensis
Frequently Bought Together
VWR®, Pipette sierologiche, Standard Line
Supplier: VWR Collection
Provette per centrifuga, Falcon®
Supplier: Corning
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
Provette per centrifuga
Supplier: Brand
VWR®, Provette e strip per PCR, 0,2 ml
Supplier: VWR Collection
VWR®, Contenitori per campioni dritti con tappo a vite
Supplier: VWR Collection
VWR®, Provette per centrifuga, senza metalli, con tappo a vite
Supplier: VWR Collection
VWR®, Pipette Pasteur, Standard Line
Supplier: VWR Collection
Cell culture flasks, Nunc™
Supplier: Thermo Fisher Scientific
VWR®, Cucchiai per campionamento
Supplier: VWR Collection
VWR®, pipette di trasferimento, alte prestazioni
Supplier: VWR Collection
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
Supplier: EPPENDORF
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF