VWR®, Macro Pipette Tips
Macro pipette tips, 5000 - 10000 µl, +, +, 160,8 mm, Clear, VWR, Eppendorf, Gilson, Rainin Classic, Biohit, and other universal-style pipettors, Rack of 25 tips
Nome del fornitore: VWR Collection
Vai a
Product Details & Documents
Improved accuracy over serological pipettes.
- Tip length facilitates access into bottles, cell culture flasks, and tubes
Universal fit for compatibility with most major pipettor brands.
: Certified free of detectable RNase, DNase, DNA, PCR inhibitors, and tested pyrogen-free. Sterile product is certified pyrogen-free.
Specifiche
- Confezione:Rack of 25 tips
- Sterili:+
- Volume:5000 - 10000 µl
- Lunghezza:160,8 mm
- Seq:12
- Stile punta:Macro pipette tips
- Colore puntale:Clear
- Per uso con:VWR, Eppendorf, Gilson, Rainin Classic, Biohit, and other universal-style pipettors
- Filtro:+